3-Amino-5-methoxypyridine ≥95%

Supplier: Matrix Scientific

057365-1G 057365-5G
101913-054EA 250.77 USD
101913-054 102551-472
3-Amino-5-methoxypyridine ≥95%
3-Amino-5-methoxypyridine

Hazard: Irritant

Formula: C₆H₈N₂O
MDL Number: MFCD09701369
CAS Number: 64436-92-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR