4-Chloro-7-fluoroquinazoline ≥95%

Supplier: Matrix Scientific

057322-1G 057322-250MG
101912-982EA 69.23 USD
101912-982 102551-464
4-Chloro-7-fluoroquinazoline ≥95%
4-Chloro-7-fluoroquinazoline

Hazard: Irritant

Formula: C₈H₄ClFN₂
MW: 182.5800 g/mol
Boiling Pt: 284 °C
Melting Pt: 131 °C
Density: 1.447
Flash Pt: 126 °C
MDL Number: MFCD08236729
CAS Number: 16499-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR