2,6-Diazaspiro[3.4]octane-6-carboxylic acid tert-butyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: 2,6-Diazaspiro[3.4]octane, N6-BOC protected, tert-Butyl 2,6-diazaspiro[3.4]octane-6-carboxylate

057301-1G 057301-250MG
101912-938EA 838.46 USD
101912-938 102551-462
2,6-Diazaspiro[3.4]octane-6-carboxylic acid tert-butyl ester ≥95%
2,6-Diazaspiro[3.4]octane-6-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₁H₂₀N₂O₂
MW: 212.29 g/mol
MDL Number: MFCD08234738
CAS Number: 885270-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR