(S)-1-Boc-(3-hydroxymethyl)pyrrolidine ≥96%

Supplier: Matrix Scientific

Synonyms: tert-butyl (3S)-3-(hydroxymethyl)pyrrolidine-1-carboxylate, (S)-1-Boc-(3-Hydroxymethyl)pyrrolidine

057265-1G 057265-5G
101912-862EA 307.69 USD
101912-862 102833-202
(S)-1-Boc-(3-hydroxymethyl)pyrrolidine ≥96%
(S)-1-Boc-(3-hydroxymethyl)pyrrolidine

Hazard: Irritant

Formula: C₁₀H₁₉NO₃
MDL Number: MFCD03094726
CAS Number: 199174-24-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR