2,6-Diazaspiro[3.5]nonane-2-carboxylic acid tert-butyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 2,6-diazaspiro[3.5]nonane-2-carboxylate

057239-1G 057239-250MG
101912-818EA 2692.31 USD
101912-818 102551-454
2,6-Diazaspiro[3.5]nonane-2-carboxylic acid tert-butyl ester ≥95%
2,6-Diazaspiro[3.5]nonane-2-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₂H₂₂N₂O₂
MW: 226.32 g/mol
MDL Number: MFCD11053736
CAS Number: 1086394-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR