4-Ethylbenzyl thiocyanate ≥98%

Supplier: Matrix Scientific

057070-10G 057070-25G 057070-100G
101912-250EA 135.38 USD
101912-250 101912-252 101912-254
4-Ethylbenzyl thiocyanate ≥98%
4-Ethylbenzyl thiocyanate

Hazard: Irritant

Formula: C₁₀H₁₁NS
MDL Number: MFCD09025690
CAS Number: 112028-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR