2,5-Diiodobenzonitrile ≥98%

Supplier: Matrix Scientific

057021-1G 057021-5G
101912-072EA 155.38 USD
101912-072 101912-074
2,5-Diiodobenzonitrile ≥98%
2,5-Diiodobenzonitrile

Hazard: Irritant

Formula: C₇H₃I₂N
MDL Number: MFCD00463830
CAS Number: 79887-24-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR