2,5-Dichlorothiobenzamide ≥98%

Supplier: Matrix Scientific

057015-1G 057015-5G
101912-052EA 141.54 USD
101912-052 101912-054
2,5-Dichlorothiobenzamide ≥98%
2,5-Dichlorothiobenzamide

Hazard: Irritant

Formula: C₇H₅Cl₂NS
MDL Number: MFCD09025686
CAS Number: 69622-81-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR