3,4-Dichlorobenzyl thiocyanate ≥98%

Supplier: Matrix Scientific

057007-10G 057007-50G
101912-022EA 73.85 USD
101912-022 101912-024
3,4-Dichlorobenzyl thiocyanate ≥98%
3,4-Dichlorobenzyl thiocyanate

Hazard: Irritant-Harmful

Formula: C₈H₅Cl₂NS
MW: 218.11 g/mol
MDL Number: MFCD09025685
CAS Number: 2082-67-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR