4-(4-Chlorophenyl)-2-butanone

Supplier: Matrix Scientific

Synonyms: 4-(4-Chlorophenyl)butan-2-one

056979-10G 056979-50G
101911-940EA 246.15 USD
101911-940 101911-942
4-(4-Chlorophenyl)-2-butanone
4-(4-Chlorophenyl)-2-butanone

Hazard: Irritant

Formula: C₁₀H₁₁ClO
MW: 182.65 g/mol
MDL Number: MFCD04004150
CAS Number: 3506-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR