3-Chloro-2-methylthiobenzamide ≥98%

Supplier: Matrix Scientific

056971-2G 056971-10G
101911-908EA 136.92 USD
101911-908 101911-910
3-Chloro-2-methylthiobenzamide ≥98%
3-Chloro-2-methylthiobenzamide

Hazard: Irritant

Formula: C₈H₈CINS
MDL Number: MFCD09025679
CAS Number: 1174906-85-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR