2-Chloro-3-methylbenzonitrile ≥98%

Supplier: Matrix Scientific

056966-1G 056966-5G
101911-900EA 135.38 USD
101911-900 101911-902
2-Chloro-3-methylbenzonitrile ≥98%
2-Chloro-3-methylbenzonitrile

Hazard: Irritant

Formula: C₈H₆CIN
MW: 151.5929 g/mol
MDL Number: MFCD09025676
CAS Number: 15013-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR