4-Chloro-3-iodobenzonitrile ≥98%

Supplier: Matrix Scientific

056957-1G 056957-5G
101911-868EA 84.62 USD
101911-868 101911-870
4-Chloro-3-iodobenzonitrile ≥98%
4-Chloro-3-iodobenzonitrile

Hazard: Irritant

Formula: C₇H₃ClIN
MDL Number: MFCD09025674
CAS Number: 914106-26-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR