4-Chloro-2-iodobenzonitrile ≥98%

Supplier: Matrix Scientific

056956-1G 056956-5G
101911-864EA 93.85 USD
101911-864 101911-866
4-Chloro-2-iodobenzonitrile ≥98%
4-Chloro-2-iodobenzonitrile

Hazard: Irritant

Formula: C₇H₃ClIN
MDL Number: MFCD09025673
CAS Number: 61272-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR