2-Chloro-5-iodobenzonitrile ≥98%

Supplier: Matrix Scientific

056955-1G 056955-5G
101911-860EA 15.38 USD
101911-860 101911-862
2-Chloro-5-iodobenzonitrile ≥98%
2-Chloro-5-iodobenzonitrile

Hazard: Irritant

Formula: C₇H₃ClIN
MDL Number: MFCD00672973
CAS Number: 289039-29-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR