3-Chlorobenzyl thiocyanate ≥98%

Supplier: Matrix Scientific

056948-10G 056948-50G
101911-832EA 160 USD
101911-832 101911-834
3-Chlorobenzyl thiocyanate ≥98%
3-Chlorobenzyl thiocyanate

Hazard: Irritant-Harmful

Formula: C₈H₆ClNS
MW: 183.66 g/mol
MDL Number: MFCD09025672
CAS Number: 64215-09-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR