1-(4-Bromophenyl)pyrrole ≥98%

Supplier: Matrix Scientific

Synonyms: 1-(4-Bromophenyl)-1H-pyrrole

056930-1G 056930-5G
101911-772EA 141.54 USD
101911-772 101911-774
1-(4-Bromophenyl)pyrrole ≥98%
1-(4-Bromophenyl)pyrrole

Hazard: Irritant

Formula: C₁₀H₈BrN
MW: 222.09 g/mol
Melting Pt: 94…96 °C
MDL Number: MFCD00128296
CAS Number: 5044-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR