4-Bromo-2-methylthiobenzamide ≥98%

Supplier: Matrix Scientific

056927-2G 056927-10G
101911-760EA 164.62 USD
101911-760 101911-762
4-Bromo-2-methylthiobenzamide ≥98%
4-Bromo-2-methylthiobenzamide

Hazard: Irritant

Formula: C₈H₈BrNS
MW: 230.12 g/mol
MDL Number: MFCD09025669
CAS Number: 397845-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR