2-Bromo-5-iodobenzonitrile ≥98%

Supplier: Matrix Scientific

056915-1G 056915-5G
101911-712EA 226.15 USD
101911-712 101911-714
2-Bromo-5-iodobenzonitrile ≥98%
2-Bromo-5-iodobenzonitrile

Hazard: Irritant

Formula: C₇H₃BrIN
Melting Pt: 144…146 °C
MDL Number: MFCD09025663
CAS Number: 1252046-13-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR