3-Bromo-4-chlorobenzonitrile ≥98%

Supplier: Matrix Scientific

056909-1G 056909-5G 056909-25G
101911-684EA 315.38 USD
101911-684 101911-686 101911-688
3-Bromo-4-chlorobenzonitrile ≥98%
3-Bromo-4-chlorobenzonitrile

Hazard: Irritant

Formula: C₇H₃BrClN
MW: 216.46 g/mol
MDL Number: MFCD09025661
CAS Number: 948549-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR