6-Bromo-3-chloro-2-cyanopyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 6-Bromo-3-chloropicolinonitrile

056857-1G 056857-5G
101911-544EA 144.62 USD
101911-544 101911-546
6-Bromo-3-chloro-2-cyanopyridine ≥98%
6-Bromo-3-chloro-2-cyanopyridine

Hazard: Irritant

Formula: C₆H₂BrClN₂
MW: 217.46 g/mol
Melting Pt: 108…110 °C
MDL Number: MFCD17014673
CAS Number: 1252046-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR