4-Butylthiobenzamide ≥98%

Supplier: Matrix Scientific

056851-1G 056851-5G
101911-534EA 184.62 USD
101911-534 101911-536
4-Butylthiobenzamide ≥98%
4-Butylthiobenzamide

Hazard: Irritant

Formula: C₁₁H₁₅NS
MDL Number: MFCD09025670
CAS Number: 1208077-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR