2-Bromo-4-methylthiobenzamide ≥98%
Supplier: Matrix Scientific
056850-1G
056850-5G
101911-530EA
103.08
USD
101911-530
101911-532
2-Bromo-4-methylthiobenzamide ≥98%
2-Bromo-4-methylthiobenzamide
Hazard: Irritant
Formula:
C₈H₈BrNS |
MDL Number:
MFCD09025668 CAS Number: 1208076-70-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...