trans-2-{3-[(tert-Butoxy)carbonyl]amino}cyclohexyl]acetic acid ≥95%

Supplier: Matrix Scientific

056515-500MG 056515-1G
101910-812EA 921.54 USD
101910-812 101910-814
trans-2-{3-[(tert-Butoxy)carbonyl]amino}cyclohexyl]acetic acid ≥95%
trans-2-{3-[(tert-Butoxy)carbonyl]amino}cyclohexyl]acetic acid

Hazard: Irritant

Formula: C₁₃H₂₃NO₄
MDL Number: MFCD16890154
CAS Number: 1242268-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR