1-tert-Butyl-3-methyl-2-oxopyrrolidine-1,3-dicarboxylate ≥95%

Supplier: Matrix Scientific

056514-500MG 056514-1G
101910-808EA 353.85 USD
101910-808 101910-810
1-tert-Butyl-3-methyl-2-oxopyrrolidine-1,3-dicarboxylate ≥95%
1-tert-Butyl-3-methyl-2-oxopyrrolidine-1,3-dicarboxylate

Hazard: Irritant

Formula: C₁₁H₁₇NO₅
MDL Number: MFCD16890151
CAS Number: 431079-79-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR