2-{[(tert-Butoxy)carbonyl]amino}-3-(pyridin-2-yl)propanoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: Boc-DL-3-(2-Pyridyl)-Ala-OH, 2-((tert-Butoxycarbonyl)amino)-3-(pyridin-2-yl)propanoic acid

056510-500MG 056510-1G
101910-792EA 453.85 USD
101910-792 101910-794
2-{[(tert-Butoxy)carbonyl]amino}-3-(pyridin-2-yl)propanoic acid ≥95%
2-{[(tert-Butoxy)carbonyl]amino}-3-(pyridin-2-yl)propanoic acid

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₄
MDL Number: MFCD02094442
CAS Number: 119434-71-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR