2-Methyl-2H-1,2,3,4-tetrazole-5-thiol ≥95%

Supplier: Matrix Scientific

056505-500MG 056505-1G
101910-770EA 1109.23 USD
101910-770 101910-772
2-Methyl-2H-1,2,3,4-tetrazole-5-thiol ≥95%
2-Methyl-2H-1,2,3,4-tetrazole-5-thiol

Hazard: Irritant

Formula: C₂H₄N₄S
MDL Number: MFCD00053366
CAS Number: 42150-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR