(2R)-3-(Ethyldisulfanyl)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid ≥95%

Supplier: Matrix Scientific

056489-500MG 056489-1G
101910-708EA 1109.23 USD
101910-708 101910-710
(2R)-3-(Ethyldisulfanyl)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid ≥95%
(2R)-3-(Ethyldisulfanyl)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid

Hazard: Irritant

Formula: C₂₀H₂₁NO₄S₂
MDL Number: MFCD00144328
CAS Number: 1208242-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR