Ethyl-3-[5-(benzyloxy)-1H-indol-3-yl]-2-{[(tert-butoxy)carbonyl]amino}propanoate ≥95%

Supplier: Matrix Scientific

056486-500MG 056486-1G 056486-5G
101910-690EA 747.69 USD
101910-690 101910-692 101910-694
Ethyl-3-[5-(benzyloxy)-1H-indol-3-yl]-2-{[(tert-butoxy)carbonyl]amino}propanoate ≥95%
Ethyl-3-[5-(benzyloxy)-1H-indol-3-yl]-2-{[(tert-butoxy)carbonyl]amino}propanoate

Hazard: Irritant

Formula: C₂₅H₃₀N₂O₅
MDL Number: MFCD16890130
CAS Number: 913718-99-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR