Ethyl-1-(3-aminopyridin-2-yl)piperidine-3-carboxylate ≥95%

Supplier: Matrix Scientific

056485-500MG 056485-1G
101910-686EA 375.38 USD
101910-686 101910-688
Ethyl-1-(3-aminopyridin-2-yl)piperidine-3-carboxylate ≥95%
Ethyl-1-(3-aminopyridin-2-yl)piperidine-3-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₉N₃O₂
MW: 249.32 g/mol
MDL Number: MFCD16890150
CAS Number: 1242267-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR