(2S)-3-[1-(2,4-Dinitrophenyl)-1H-imidazol-4-yl]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic ≥95%

Supplier: Matrix Scientific

Synonyms: Fmoc-His(DNP)-OH, 1-(2,4-Dinitrophenyl)-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-histidine

056482-500MG 056482-1G
101910-674EA 1109.23 USD
101910-674 101910-676
(2S)-3-[1-(2,4-Dinitrophenyl)-1H-imidazol-4-yl]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic ≥95%
(2S)-3-[1-(2,4-Dinitrophenyl)-1H-imidazol-4-yl]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic

Hazard: Irritant

Formula: C₂₇H₂₁N₅O₈
MW: 543.5 g/mol
MDL Number: MFCD00077060
CAS Number: 83999-94-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR