2,3-Dihydro-1,4-benzodioxin-6-ylmethanol ≥95%

Supplier: Matrix Scientific

Synonyms: 2,3-Dihydro-6-(hydroxymethyl)-1,4-benzodioxine, (2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)methanol

056478-500MG 056478-1G
101910-656EA 115.38 USD
101910-656 101910-658
2,3-Dihydro-1,4-benzodioxin-6-ylmethanol ≥95%
2,3-Dihydro-1,4-benzodioxin-6-ylmethanol

Hazard: Irritant

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
MDL Number: MFCD01317581
CAS Number: 39270-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR