J-502380 ≥95%

Supplier: Matrix Scientific

Synonyms: Fmoc-Glu(OSu)-OtBu

056466-500MG 056466-1G
101910-608EA 556.92 USD
101910-608 101910-610
J-502380 ≥95%
J-502380

Hazard: Irritant

Formula: C₂₈H₃₀N₂O₈
MW: 522.554 g/mol
MDL Number: MFCD00237021
CAS Number: 200616-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR