4,5,6,7-Tetrahydro-1H-benzimidazole-5-carboxylic acid ≥95%

Supplier: Matrix Scientific

056462-500MG 056462-1G
101910-594EA 81.54 USD
101910-594 101910-596
4,5,6,7-Tetrahydro-1H-benzimidazole-5-carboxylic acid ≥95%
4,5,6,7-Tetrahydro-1H-benzimidazole-5-carboxylic acid

Hazard: Irritant

Formula: C₈H₁₀N₂O₂
MDL Number: MFCD03839866
CAS Number: 26751-24-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR