tert-Butyl piperidine-3-carboxylate hydrochloride ≥95%

Supplier: Matrix Scientific

056460-500MG 056460-1G
101910-586EA 556.92 USD
101910-586 101910-588
tert-Butyl piperidine-3-carboxylate hydrochloride ≥95%
tert-Butyl piperidine-3-carboxylate hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₀ClNO₂
MDL Number: MFCD04038695
CAS Number: 301180-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR