4-(2-((tert-Butoxycarbonyl)amino)ethyl)benzoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(2-Boc-aminoethyl)benzoic acid

056447-500MG 056447-1G
101910-538EA 375.38 USD
101910-538 101910-540
4-(2-((tert-Butoxycarbonyl)amino)ethyl)benzoic acid ≥95%
4-(2-((tert-Butoxycarbonyl)amino)ethyl)benzoic acid

Hazard: Irritant

Formula: C₁₄H₁₉NO₄
MW: 265.3050 g/mol
MDL Number: MFCD07371516
CAS Number: 132690-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR