2-{[(tert-Butoxy)carbonyl]amino}-3-(pyridin-4-yl)propanoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-((tert-Butoxycarbonyl)amino)-3-(pyridin-4-yl)propanoic acid

056446-500MG 056446-1G
101910-534EA 484.62 USD
101910-534 101910-536
2-{[(tert-Butoxy)carbonyl]amino}-3-(pyridin-4-yl)propanoic acid ≥95%
2-{[(tert-Butoxy)carbonyl]amino}-3-(pyridin-4-yl)propanoic acid

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₄
MW: 266.3 g/mol
MDL Number: MFCD06656857
CAS Number: 33814-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR