3-(Pyridin-4-yl)propan-1-amine ≥95%

Supplier: Matrix Scientific

056443-500MG 056443-1G
101910-522EA 384.62 USD
101910-522 101910-524
3-(Pyridin-4-yl)propan-1-amine ≥95%
3-(Pyridin-4-yl)propan-1-amine

Hazard: Irritant

Formula: C₈H₁₂N₂
MDL Number: MFCD08234495
CAS Number: 30532-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR