Methyl-3-(trifluoromethyl)pyridine-4-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-(trifluoromethyl)isonicotinate

056440-500MG 056440-1G
101910-510EA 453.85 USD
101910-510 101910-512
Methyl-3-(trifluoromethyl)pyridine-4-carboxylate ≥95%
Methyl-3-(trifluoromethyl)pyridine-4-carboxylate

Hazard: Irritant

Formula: C₈H₆F₃NO₂
MW: 205.14 g/mol
MDL Number: MFCD14698095
CAS Number: 1203952-88-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR