1-(Methylsulfonyl)spiro[indoline-3,4'-piperidine]hydrochloride ≥95%

Supplier: Matrix Scientific

056437-500MG 056437-1G
101910-496EA 747.69 USD
101910-496 101910-498
1-(Methylsulfonyl)spiro[indoline-3,4'-piperidine]hydrochloride ≥95%
1-(Methylsulfonyl)spiro[indoline-3,4'-piperidine]hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂O₂S
MDL Number: MFCD09033869
CAS Number: 159634-86-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR