1-[2-(Methylsulfanyl)phenyl]piperazine ≥95%

Supplier: Matrix Scientific

Synonyms: 1-[2-(Methylthio)phenyl]piperazine

056436-500MG 056436-1G
101910-492EA 181.54 USD
101910-492 101910-494
1-[2-(Methylsulfanyl)phenyl]piperazine ≥95%
1-[2-(Methylsulfanyl)phenyl]piperazine

Hazard: Irritant/Stored under Argon

Formula: C₁₁H₁₆N₂S
MW: 208.33 g/mol
MDL Number: MFCD00040795
CAS Number: 1013-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR