Methyl-5-bromo-2-chloropyridine-4-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 5-bromo-2-chloroisonicotinate

056425-500MG 056425-1G 056425-5G
101910-456EA 123.08 USD
101910-456 101910-458 101910-460
Methyl-5-bromo-2-chloropyridine-4-carboxylate ≥95%
Methyl-5-bromo-2-chloropyridine-4-carboxylate

Hazard: Irritant

Formula: C₇H₅BrClNO₂
MW: 250.49 g/mol
MDL Number: MFCD07375113
CAS Number: 886365-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR