Methyl-5-amino-2-[2-(propan-2-yl)piperazin-1-yl]benzoate ≥95%

Supplier: Matrix Scientific

056419-500MG 056419-1G
101910-438EA 1670.77 USD
101910-438 101910-440
Methyl-5-amino-2-[2-(propan-2-yl)piperazin-1-yl]benzoate ≥95%
Methyl-5-amino-2-[2-(propan-2-yl)piperazin-1-yl]benzoate

Hazard: Irritant

Formula: C₁₅H₂₃N₃O₂
MDL Number: MFCD16710283
CAS Number: 1242268-13-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR