N-[2-(4-Hydroxyphenyl)ethyl]methanesulfonamide ≥95%

Supplier: Matrix Scientific

056409-500MG 056409-1G
101910-392EA 556.92 USD
101910-392 101910-394
N-[2-(4-Hydroxyphenyl)ethyl]methanesulfonamide ≥95%
N-[2-(4-Hydroxyphenyl)ethyl]methanesulfonamide

Hazard: Irritant

Formula: C₉H₁₃NO₃S
MDL Number: MFCD02682911
CAS Number: 883107-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR