9H-Fluoren-9-ylmethyl-4-aminopiperidine-1-carboxylate hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: (9H-Fluoren-9-yl)methyl 4-aminopiperidine-1-carboxylate hydrochloride

056406-500MG 056406-1G
101910-380EA 181.54 USD
101910-380 101910-382
9H-Fluoren-9-ylmethyl-4-aminopiperidine-1-carboxylate hydrochloride ≥95%
9H-Fluoren-9-ylmethyl-4-aminopiperidine-1-carboxylate hydrochloride

Hazard: Irritant

Formula: C₂₀H₂₃ClN₂O₂
MW: 358.87 g/mol
MDL Number: MFCD03001675
CAS Number: 811841-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR