9H-Fluoren-9-ylmethyl 4-(aminomethyl)piperidine-1-carboxylate hydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
(9H-Fluoren-9-yl)methyl 4-(aminomethyl)piperidine-1-carboxylate hydrochloride
056405-500MG
056405-1G
101910-376EA
330.77
USD
101910-376
101910-378
9H-Fluoren-9-ylmethyl 4-(aminomethyl)piperidine-1-carboxylate hydrochloride ≥95%
9H-Fluoren-9-ylmethyl 4-(aminomethyl)piperidine-1-carboxylate hydrochloride
Hazard: Irritant
Formula:
C₂₁H₂₅ClN₂O₂ MW: 372.9 g/mol |
MDL Number:
MFCD02179398 CAS Number: 391248-14-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...