3-Ethyl-1-[2-(4-hydroxyphenyl)ethyl]urea ≥95%

Supplier: Matrix Scientific

056403-500MG 056403-1G 056403-5G
101910-366EA 556.92 USD
101910-366 101910-368 101910-370
3-Ethyl-1-[2-(4-hydroxyphenyl)ethyl]urea ≥95%
3-Ethyl-1-[2-(4-hydroxyphenyl)ethyl]urea

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₂
MDL Number: MFCD02682910
CAS Number: 883107-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR