N-Ethyl-4-(2-hydroxyethyl)piperazine-1-carboxamide ≥95%

Supplier: Matrix Scientific

056401-500MG 056401-1G
101910-358EA 556.92 USD
101910-358 101910-360
N-Ethyl-4-(2-hydroxyethyl)piperazine-1-carboxamide ≥95%
N-Ethyl-4-(2-hydroxyethyl)piperazine-1-carboxamide

Hazard: Irritant

Formula: C₉H₁₉N₃O₂
MDL Number: MFCD06410746
CAS Number: 816456-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR