N-[2-(Dimethylamino)ethyl]piperidine-4-carboxamide ≥95%

Supplier: Matrix Scientific

056396-500MG 056396-1G
101910-338EA 921.54 USD
101910-338 101910-340
N-[2-(Dimethylamino)ethyl]piperidine-4-carboxamide ≥95%
N-[2-(Dimethylamino)ethyl]piperidine-4-carboxamide

Hazard: Irritant

Formula: C₁₀H₂₁N₃O
MDL Number: MFCD06739721
CAS Number: 886504-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR