7,8-Dimethoxy-4,5-dihydrospiro(pyrrolo[1,2-a]quinoxaline-4,4'-piperidine) hydrochloride ≥95%

Supplier: Matrix Scientific

056394-500MG 056394-1G
101910-330EA 1109.23 USD
101910-330 101910-332
7,8-Dimethoxy-4,5-dihydrospiro(pyrrolo[1,2-a]quinoxaline-4,4'-piperidine) hydrochloride ≥95%
7,8-Dimethoxy-4,5-dihydrospiro(pyrrolo[1,2-a]quinoxaline-4,4'-piperidine) hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₂ClN₃O₂
MDL Number: MFCD16710287
CAS Number: 1242268-29-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR